accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNSNHOOOOCH3

CAS RN 1421262-42-1

IUPAC Name: 2-[2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-methyl-1,3-thiazol-4-yl]phenyl acetate
Synonyms: 2-(2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-methyl-1,3-thiazol-4-yl)phenyl acetate
Molecular Weight (g/mol): 470.54
Molecular Formula: C27H22N2O4S
InChi Key: MAUMOHZLUIAQJC-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC=C1C1=C(C)SC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results