accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOSONHN

CAS RN 1421262-49-8

IUPAC Name: 2-{4-oxo-3H,4H-thieno[2,3-d]pyrimidin-5-yl}phenyl acetate
Synonyms: 2-{4-oxo-3H-thieno[2,3-d]pyrimidin-5-yl}phenyl acetate
Molecular Weight (g/mol): 286.31
Molecular Formula: C14H10N2O3S
InChi Key: FKFWGMGSNRHSIW-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC=C1C1=CSC2=C1C(=O)NC=N2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results