accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSOOBr

CAS RN 1421262-56-7

IUPAC Name: ethyl 3-bromobenzene-1-sulfonate
Synonyms: ethyl 3-bromobenzenesulfonate
Molecular Weight (g/mol): 265.12
Molecular Formula: C8H9BrO3S
InChi Key: KVPGXLXCDWOMIC-UHFFFAOYSA-N
SMILES: CCOS(=O)(=O)C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results