accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNOO

CAS RN 1421262-92-1

IUPAC Name: 3-[(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]pyridine-2-carbonitrile
Synonyms: 3-[(2,5-dioxopyrrol-1-yl)methyl]pyridine-2-carbonitrile
Molecular Weight (g/mol): 213.2
Molecular Formula: C11H7N3O2
InChi Key: XGKQSALDRPJQPL-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1CC1=C(N=CC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results