accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSONHN

CAS RN 1421263-78-6

IUPAC Name: 5-(pyridin-4-yl)-3H,4H-thieno[2,3-d]pyrimidin-4-one
Synonyms: 5-(pyridin-4-yl)-3H-thieno[2,3-d]pyrimidin-4-one
Molecular Weight (g/mol): 229.26
Molecular Formula: C11H7N3OS
InChi Key: DUWHOSMEYMIDKJ-UHFFFAOYSA-N
SMILES: O=C1NC=NC2=C1C(=CS2)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results