accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBr

CAS RN 142137-17-5

IUPAC Name: 3-bromo-5-phenylpyridine
Synonyms: 3-bromo-5-phenylpyridine
Molecular Weight (g/mol): 234.1
Molecular Formula: C11H8BrN
InChi Key: AWCQJXPOCRXHNK-UHFFFAOYSA-N
SMILES: BrC1=CN=CC(=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results