accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142404-82-8

IUPAC Name: N-(5-bromo-4-methylpyridin-2-yl)acetamide
Synonyms: N-(5-bromo-4-methylpyridin-2-yl)acetamide
Molecular Weight (g/mol): 229.08
Molecular Formula: C8H9BrN2O
InChi Key: WVJCOPBDWMWUNP-UHFFFAOYSA-N
SMILES: CC(=O)NC1=NC=C(Br)C(C)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results