accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3NONHH3CCH3

CAS RN 14252-80-3

IUPAC Name: hydrogen 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide chloride
Synonyms: hydrogen bupivacaine chloride
Molecular Weight (g/mol): 324.89
Molecular Formula: C18H29ClN2O
InChi Key: SIEYLFHKZGLBNX-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results