CAS RN 1426138-42-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ FX 1
Bcl-6 inhibitor; 10-fold more potent than endogenous Bcl-6 corepressors