Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ FX 1
Description
FX 1 is a Bcl-6 inhibitor (Kd = 3 μM); exhibits 10-fold greater potency than endogenous Bcl-6 corepressors. Disrupts formation of Bcl-6 repression complex. Induces regression of established tumors in mice bearing DLBCL xenograft.
Specifications
Specifications
| Chemical Name or Material | (5Z)-5-(5-Chloro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)-4-oxo-2-thioxo-3-thiazolidinepropanoic acid |
| CAS | 1426138-42-2 |
| Quantity | 5 mg |
| Molecular Formula | C14H9ClN2O4S2 |
| Formula Weight | 368.82 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?