accessibility menu, dialog, popup

UserName

Viewing profile as user:
FePPClClClCl

CAS RN 142691-70-1

IUPAC Name: 1,1'-Bis(dichlorophosphino)ferrocene
Synonyms: —
Molecular Weight (g/mol): 387.77
Molecular Formula: C10H8Cl4FeP2
InChi Key: —
SMILES: [Fe].ClP(Cl)c1cccc1.ClP(Cl)c1cccc1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)