Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bis(dichlorophosphino)ferrocene, 98%, Thermo Scientific Chemicals

Catalog No. AAH27293MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27293MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH27293MD Supplier Thermo Scientific Chemicals Supplier No. H27293MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

It is applied as a reactant for synthesis of ferrocene-based bidentate phosphonite ligands for rhodium(I)-catalyzed enantioselective hydroformylation, metallocene-bridged bisphosphaalkene and as a catalyst for synthesis of oligonuclear heterometallic complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a reactant for synthesis of ferrocene-based bidentate phosphonite ligands for rhodium(I)-catalyzed enantioselective hydroformylation, metallocene-bridged bisphosphaalkene and as a catalyst for synthesis of oligonuclear heterometallic complexes.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from water/moisture, strong bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142691-70-1
Molecular Formula C10H8Cl4FeP2
Molecular Weight (g/mol) 387.77
MDL Number MFCD01630854
IUPAC Name 1,1'-Bis(dichlorophosphino)ferrocene
SMILES [Fe].ClP(Cl)c1cccc1.ClP(Cl)c1cccc1

Specifications

Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 387.78
Percent Purity 98%
Chemical Name or Material 1,1'-Bis(dichlorophosphino)ferrocene
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.