accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1427397-15-6

IUPAC Name: methyl 2-(2-bromo-6-fluorophenyl)acetate
Synonyms: methyl 2-(2-bromo-6-fluorophenyl)acetate
Molecular Weight (g/mol): 247.06
Molecular Formula: C9H8BrFO2
InChi Key: QXIUUACWTNNJID-UHFFFAOYSA-N
SMILES: COC(=O)CC1=C(F)C=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results