accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHO(S)HNNHO(R)H3CCH3OHNOH

CAS RN 142880-36-2

IUPAC Name: (2R)-N'-hydroxy-N-[(1S)-2-(1H-indol-3-yl)-1-(methylcarbamoyl)ethyl]-2-(2-methylpropyl)butanediamide
Synonyms: ilomastat
Molecular Weight (g/mol): 388.47
Molecular Formula: C20H28N4O4
InChi Key: NITYDPDXAAFEIT-DYVFJYSZSA-N
SMILES: CNC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)[C@H](CC(C)C)CC(=O)NO
Form 
  • (2)
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)