CAS RN 143-33-9
Sodium cyanide, granules, 98%, Thermo Scientific Chemicals
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium;cyanide SMILES: [Na+].[C-]#N
Sodium cyanide, 98+%, extra pure, Thermo Scientific Chemicals
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 SMILES: [Na+].[C-]#N
Sodium cyanide, ACS, 95% min, Thermo Scientific Chemicals
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium;cyanide SMILES: [Na+].[C-]#N
Sodium Cyanide, Reagent, ACS, 95%, Spectrum™ Chemical
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N IUPAC Name: sodium iminomethanide SMILES: [Na+].[C-]#N
Sodium cyanide, 98%, for analysis, Thermo Scientific Chemicals
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium iminomethanide SMILES: [Na+].[C-]#N
Sodium Cyanide, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium iminomethanide SMILES: [Na+].[C-]#N
Sodium Cyanide, ACS Reagent Grade, Ricca Chemical
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N IUPAC Name: sodium iminomethanide SMILES: [Na+].[C-]#N
Sodium Cyanide Standard, 500 ppm CN-, Ricca Chemical
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N PubChem CID: 8929 IUPAC Name: sodium iminomethanide SMILES: [Na+].[C-]#N
Sodium Cyanide, 2.5% (w/v) (25 g/L), Ricca Chemical
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium iminomethanide SMILES: [Na+].[C-]#N
Sodium Cyanide (Granular/Certified ACS), Fisher Chemical™
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium iminomethanide SMILES: [Na+].[C-]#N
Sodium Cyanide (Granular/Technical), Fisher Chemical™
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium iminomethanide SMILES: [Na+].[C-]#N
Sodium cyanide, 95+%, ACS reagent, Thermo Scientific Chemicals
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 SMILES: [Na+].[C-]#N
Sodium Cyanide, ≥95%, (ACS Reagent Grade), MP Biomedicals
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium iminomethanide SMILES: [Na+].[C-]#N
Avantor Macron Sodium Cyanide ACS AR Granular, Macron™
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium iminomethanide SMILES: [Na+].[C-]#N
Sodium Cyanide, ACS Reagent, ≥95.0%, Solstice
CAS: 143-33-9 Molecular Formula: CNNa Molecular Weight (g/mol): 49.01 MDL Number: MFCD00003523 InChI Key: MNWBNISUBARLIT-UHFFFAOYSA-N Synonym: sodium cyanide,cyanogran,cymag,cyanide of sodium,sodium cyanide na cn,nacn,cyanobrik,cyanide salts,kyanid sodny,cyanasalt h PubChem CID: 8929 IUPAC Name: sodium;cyanide SMILES: [Na+].[C-]#N