Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium cyanide, 95+%, ACS reagent, Thermo Scientific Chemicals

Catalog No. AC424305000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424305000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC424305000 Supplier Thermo Scientific Chemicals Supplier No. 424305000

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
Synonym sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
PubChem CID 8929
SMILES [Na+].[C-]#N

Specifications

Color White
Physical Form Crystalline Powder, Flakes, and Chunks
Quantity 500 g
Assay Percent Range 95+%
Linear Formula NaCN
Beilstein 02, IV, 50
Fieser 04,446; 05,506; 06,535; 07,333; 08,453; 11,481; 13,185
Merck Index 15, 8741
IUPAC Name sodium;cyanide
Formula Weight 49
Percent Purity ≥95.0%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Sodium cyanide, 95+%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May be corrosive to metals. Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects. Contact with acids liberates very toxic gas.
Precautionary Statement GHS P Statement
Absorb spillage to prevent material damage. Wear protective gloves/protective clothing. Rinse mouth. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 205-599-4
RTECSNumber VZ7525000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.