Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Cyanide (Granular/Technical), Fisher Chemical™

Catalog No. S283I500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S283I-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S283I-500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
Synonym sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
PubChem CID 8929
IUPAC Name sodium iminomethanide
SMILES [Na+].[C-]#N

Specifications

Boiling Point 1497°C
Melting Point 562°C
Color White
pH 11 to 12
Physical Form Solid
Quantity 500 g
Formula Weight 49.01
Grade Technical
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 1hPa at 817°C
Density 1.6000g/cm³
Chemical Name or Material Sodium Cyanide
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 6.1, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.