CAS RN 1430844-80-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ A 1331852
Potent and selective Bcl-xL inhibitor; antitumor and inducer of apoptosis