CAS RN 143086-32-2
IUPAC Name:
N-(4-phenylbutan-2-yl)pentanamide
Synonyms:
N-(4-phenylbutan-2-yl)pentanamide
Molecular Weight (g/mol):
233.36
Molecular Formula:
C15H23NO
InChi Key:
PLWKOZGQPUWQNJ-UHFFFAOYNA-N
SMILES:
CCCCC(=O)NC(C)CCC1=CC=CC=C1
1
–
1
of
1
results
1
N-(1-Methyl-3-phenylpropyl)valeramide, TCI America™
CAS: 143086-32-2 Molecular Formula: C15H23NO Molecular Weight (g/mol): 233.355 MDL Number: MFCD00144821 InChI Key: PLWKOZGQPUWQNJ-UHFFFAOYSA-N Synonym: 1-Phenyl-3-valeramidobutane PubChem CID: 4657091 IUPAC Name: N-(4-phenylbutan-2-yl)pentanamide SMILES: CCCCC(=O)NC(C)CCC1=CC=CC=C1