CAS RN 1432660-47-3
IUPAC Name:
3-[5-(cyclopropylsulfamoyl)-2-(thiophen-3-yl)phenyl]-1-[3-(trifluoromethyl)phenyl]urea
Synonyms:
3-[5-(cyclopropylsulfamoyl)-2-(thiophen-3-yl)phenyl]-1-[3-(trifluoromethyl)phenyl]urea
Molecular Weight (g/mol):
481.51
Molecular Formula:
C21H18F3N3O3S2
InChi Key:
CCAWRGNYALGPQH-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(NC(=O)NC2=C(C=CC(=C2)S(=O)(=O)NC2CC2)C2=CSC=C2)=CC=C1
1
–
2
of
2
results
2
MilliporeSigma™ Calbiochem™ IDH2 Inhibitor, R140Q Somatic Mutant-Specific, AGI-6780
A cell-permeable inhibitor of isocitrate dehydrogenase 2 R140Q mutant-containing dimers. Has minimal effect on wild type dimer-catalyzed alpha-KG production.