CAS RN 143314-14-1
CAS RN 143314-14-1
IUPAC Name:
1-ethyl-3-methyl-1H-imidazol-3-ium nitrate
Synonyms:
1-ethyl-3-methylimidazolium nitrate
Molecular Weight (g/mol):
173.17
Molecular Formula:
C6H11N3O3
InChi Key:
JDOJFSVGXRJFLL-UHFFFAOYSA-N
SMILES:
[O-][N+]([O-])=O.CCN1C=C[N+](C)=C1
1
–
1
of
1
results
1
1-Ethyl-3-methylimidazolium Nitrate 98.0+%, TCI America™
CAS: 143314-14-1 Molecular Formula: C6H11N3O3 Molecular Weight (g/mol): 173.172 MDL Number: MFCD00210007 InChI Key: JDOJFSVGXRJFLL-UHFFFAOYSA-N Synonym: 1-ethyl-3-methylimidazolium nitrate,1-ethyl-3-methylimidazolium nitrate nt,emim no3,acmc-20dtjx,dsstox_cid_29073,dsstox_rid_83292,dsstox_gsid_49217,ksc170o1h,1-ethyl-3-methyl-1h-imidazol-3-ium nitrate PubChem CID: 16211072 IUPAC Name: 1-ethyl-3-methylimidazol-3-ium;nitrate SMILES: CCN1C=C[N+](=C1)C.[N+](=O)([O-])[O-]