accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 14347-83-2

IUPAC Name: 4-[(benzyloxy)methyl]-2,2-dimethyl-1,3-dioxolane
Synonyms: 4-[(benzyloxy)methyl]-2,2-dimethyl-1,3-dioxolane
Molecular Weight (g/mol): 222.28
Molecular Formula: C13H18O3
InChi Key: DBFDSKSLTCMIPB-UHFFFAOYNA-N
SMILES: CC1(C)OCC(COCC2=CC=CC=C2)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results