accessibility menu, dialog, popup

UserName

Viewing profile as user:
FNFF

CAS RN 143879-80-5

IUPAC Name: 2,3,4-trifluorobenzonitrile
Synonyms: 2,3,4-trifluorobenzonitrile
Molecular Weight (g/mol): 157.1
Molecular Formula: C7H2F3N
InChi Key: KTPHYLJFAZNALV-UHFFFAOYSA-N
SMILES: FC1=CC=C(C#N)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results