accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14401-10-6

IUPAC Name: 2-amino-6-[(2,4-dinitrophenyl)amino]hexanoic acid hydrochloridyl
Synonyms: epsilon-dnp-lysine hydrochloridyl
Molecular Weight (g/mol): 347.73
Molecular Formula: C12H16ClN4O6
InChi Key: FJDUWQCVTQNLPA-UHFFFAOYNA-N
SMILES: [Cl].NC(CCCCNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results