accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14404-33-2

ClRuClClClClKK

CAS RN 14404-33-2

IUPAC Name: dipotassium pentachlororutheniumdiuide
Synonyms: dipotassium pentachlororutheniumdiuide
Molecular Weight (g/mol): 356.52
Molecular Formula: Cl5K2Ru
InChi Key: IXYMQZXAJLWXRK-UHFFFAOYSA-I
SMILES: [K+].[K+].Cl[Ru--](Cl)(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)