accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1443036-49-4

IUPAC Name: 1-phenyl-2-(phenyl-λ³-iodanylidene)-2-trifluoromethanesulfonylethan-1-one
Synonyms: 1-phenyl-2-(phenyl-λ³-iodanylidene)-2-trifluoromethanesulfonylethanone
Molecular Weight (g/mol): 454.2
Molecular Formula: C15H10F3IO3S
InChi Key: BGLWFUSLNKHJEU-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)C(=[I]C1=CC=CC=C1)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results