accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14448-67-0

IUPAC Name: hydrogen (2Z)-2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole chloride
Synonyms: hydrogen (2Z)-2-hydrazinylidene-3-methyl-1,3-benzothiazole chloride
Molecular Weight (g/mol): 215.7
Molecular Formula: C8H10ClN3S
InChi Key: OEZPVSPULCMUQB-DQMXGCRQSA-N
SMILES: [H+].[Cl-].CN1\C(SC2=CC=CC=C12)=N\N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results