accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNN

CAS RN 1448-87-9

IUPAC Name: 2-chloroquinoxaline
Synonyms: 2-chloroquinoxaline
Molecular Weight (g/mol): 164.59
Molecular Formula: C8H5ClN2
InChi Key: BYHVGQHIAFURIL-UHFFFAOYSA-N
SMILES: ClC1=CN=C2C=CC=CC2=N1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)