accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONBrN

CAS RN 1448636-19-8

IUPAC Name: 6-(bromomethyl)-3-cyanopyridin-2-yl acetate
Synonyms: 6-(bromomethyl)-3-cyanopyridin-2-yl acetate
Molecular Weight (g/mol): 255.07
Molecular Formula: C9H7BrN2O2
InChi Key: KRLBCAXBCMWUII-UHFFFAOYSA-N
SMILES: CC(=O)OC1=C(C=CC(CBr)=N1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results