accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14491-66-8

IUPAC Name: 1,4-dioctyl butanedioate
Synonyms: dicapryl succinate
Molecular Weight (g/mol): 342.52
Molecular Formula: C20H38O4
InChi Key: KWABLUYIOFEZOY-UHFFFAOYSA-N
SMILES: CCCCCCCCOC(=O)CCC(=O)OCCCCCCCC
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results