accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFOHNClClHOOH

CAS RN 1449132-60-8

IUPAC Name: {3-[(2,3-dichlorophenyl)carbamoyl]-5-fluorophenyl}boronic acid
Synonyms: 3-[(2,3-dichlorophenyl)carbamoyl]-5-fluorophenylboronic acid
Molecular Weight (g/mol): 327.93
Molecular Formula: C13H9BCl2FNO3
InChi Key: DEEWNQYDQBIFSS-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC(F)=C1)C(=O)NC1=C(Cl)C(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results