Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2,3-Dichlorophenylcarbamoyl)-3-fluorobenzeneboronic acid, 97%, Thermo Scientific™

Catalog No. AAH54500MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54500MD 250 mg
AAH5450003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54500MD Supplier Thermo Scientific Chemicals Supplier No. H54500MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1449132-60-8
Molecular Formula C13H9BCl2FNO3
Molecular Weight (g/mol) 327.93
MDL Number MFCD20040205
InChI Key DEEWNQYDQBIFSS-UHFFFAOYSA-N
Synonym 5-2,3-dichlorophenylcarbamoyl-3-fluorobenzeneboronic acid
PubChem CID 73995843

Specifications

Quantity 250 mg
Formula Weight 327.93
Percent Purity 97%
Chemical Name or Material 5-(2,3-Dichlorophenylcarbamoyl)-3-fluorobenzeneboronic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.