accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFOHNClNOOHOOH

CAS RN 1449133-18-9

IUPAC Name: {3-[(2-chloro-6-nitrophenyl)carbamoyl]-5-fluorophenyl}boronic acid
Synonyms: 3-[(2-chloro-6-nitrophenyl)carbamoyl]-5-fluorophenylboronic acid
Molecular Weight (g/mol): 338.48
Molecular Formula: C13H9BClFN2O5
InChi Key: SMRJKQIAHFDGKT-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC(F)=C1)C(=O)NC1=C(C=CC=C1Cl)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results