accessibility menu, dialog, popup

UserName

Viewing profile as user:
BONHFNOOFHOOH

CAS RN 1449144-94-8

IUPAC Name: {2-fluoro-4-[(4-fluoro-2-nitrophenyl)carbamoyl]phenyl}boronic acid
Synonyms: 2-fluoro-4-[(4-fluoro-2-nitrophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 322.03
Molecular Formula: C13H9BF2N2O5
InChi Key: JSRCTYHYPFPKJW-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=C(C=C1)C(=O)NC1=C(C=C(F)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results