CAS RN 145-50-6
alpha-Naphtholbenzein
CAS: 145-50-6 Molecular Formula: C27H18O2 Molecular Weight (g/mol): 374.439 MDL Number: MFCD00078492 InChI Key: VDDWRTZCUJCDJM-PNHLSOANSA-N PubChem CID: 5941340 IUPAC Name: (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one SMILES: C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O
p-Naphtholbenzein Indicator Solution, Fisher Chemical
CAS: 145-50-6 Molecular Formula: C27H18O2 Molecular Weight (g/mol): 374.439 MDL Number: MFCD00078492 InChI Key: VDDWRTZCUJCDJM-PNHLSOANSA-N PubChem CID: 5941340 IUPAC Name: (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one SMILES: C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O
p-Naphtholbenzein, TCI America™
CAS: 145-50-6 Molecular Formula: C27H18O2 Molecular Weight (g/mol): 374.439 MDL Number: MFCD00078492 InChI Key: VDDWRTZCUJCDJM-PNHLSOANSA-N Synonym: p-naphtholbenzein,1-naphtholbenzein,alpha-naphtholbenzein,.alpha.-naphtholbenzein,4-4-hydroxynaphthyl phenylmethylene naphthalen-1-one,naphtholbenzein,1 4h-naphthalenone, 4-4-hydroxy-1-naphthalenyl phenylmethylene,1 4h-naphthalenone,4-4-hydroxy-1-naphthalenyl phenylmethylene,1 4h-naphthalenone, 4-.alpha.-4-hydroxy-1-naphthyl benzylidene PubChem CID: 5941340 IUPAC Name: (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one SMILES: C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O
Alpha-Naphtholbenzein, Sigma-Aldrich
CAS: 145-50-6 Molecular Weight (g/mol): 374.43 g/mol
Thermo Scientific Chemicals alpha-Naphtholbenzein
CAS: 145-50-6 Molecular Formula: C27H18O2 Molecular Weight (g/mol): 374.44 InChI Key: VDDWRTZCUJCDJM-PNHLSOANSA-N PubChem CID: 5941340 IUPAC Name: (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one SMILES: C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O
p-Naphtholbenzein TS, 0.25% (w/v) in Glacial Acetic Acid, Ricca Chemical
CAS: 145-50-6 Molecular Formula: C27H18O2 Molecular Weight (g/mol): 374.439 InChI Key: VDDWRTZCUJCDJM-PNHLSOANSA-N PubChem CID: 5941340 IUPAC Name: (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one SMILES: C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O
p-Naphtholbenzein (Reagent), Fisher Chemical
CAS: 145-50-6 Molecular Formula: C27H18O2 Molecular Weight (g/mol): 374.439 MDL Number: MFCD00078492 InChI Key: VDDWRTZCUJCDJM-PNHLSOANSA-N Synonym: p-naphtholbenzein,1-naphtholbenzein,alpha-naphtholbenzein,.alpha.-naphtholbenzein,4-4-hydroxynaphthyl phenylmethylene naphthalen-1-one,naphtholbenzein,1 4h-naphthalenone, 4-4-hydroxy-1-naphthalenyl phenylmethylene,1 4h-naphthalenone,4-4-hydroxy-1-naphthalenyl phenylmethylene,1 4h-naphthalenone, 4-.alpha.-4-hydroxy-1-naphthyl benzylidene PubChem CID: 5941340 IUPAC Name: (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one SMILES: C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O
α-Naphtholbenzein, 98%, Thermo Scientific™
CAS: 145-50-6 Molecular Formula: C27H18O2 Molecular Weight (g/mol): 374.439 InChI Key: VDDWRTZCUJCDJM-PNHLSOANSA-N Synonym: p-naphtholbenzein,1-naphtholbenzein,alpha-naphtholbenzein,.alpha.-naphtholbenzein,4-4-hydroxynaphthyl phenylmethylene naphthalen-1-one,naphtholbenzein,1 4h-naphthalenone, 4-4-hydroxy-1-naphthalenyl phenylmethylene,1 4h-naphthalenone,4-4-hydroxy-1-naphthalenyl phenylmethylene,1 4h-naphthalenone, 4-.alpha.-4-hydroxy-1-naphthyl benzylidene PubChem CID: 5941340 IUPAC Name: (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one SMILES: C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O
Avantor Macron p-Naphtholbenzein OR, Macron Fine Chemicals™
CAS: 145-50-6 Molecular Formula: C27H18O2 Molecular Weight (g/mol): 374.439 InChI Key: VDDWRTZCUJCDJM-PNHLSOANSA-N PubChem CID: 5941340 IUPAC Name: (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one SMILES: C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O