Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha-Naphtholbenzein, Sigma-Aldrich

Catalog No. 01258934 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-934 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-934 Supplier MilliporeSigma Supelco Supplier No. 29109925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indicator grade dye suitable for pH titrations and detection of bacterial growth.

Alpha-Naphtholbenzein has been used as an indicator during sequential injection analysis technique done for acid-base titrations.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145-50-6
Molecular Weight (g/mol) 374.43 g/mol

Specifications

Color Black to Brown
pH 8.2-10
Quantity 25 g
Grade Indicator
Chemical Name or Material Alpha-Naphtholbenzein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.