Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Naphtholbenzein, 98%, Thermo Scientific™

Catalog No. AC415320100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC415320100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC415320100 Supplier Thermo Scientific Chemicals Supplier No. 415320100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145-50-6
Molecular Formula C27H18O2
Molecular Weight (g/mol) 374.439
InChI Key VDDWRTZCUJCDJM-PNHLSOANSA-N
Synonym p-naphtholbenzein, 1-naphtholbenzein, alpha-naphtholbenzein, .alpha.-naphtholbenzein, 4-4-hydroxynaphthyl phenylmethylene naphthalen-1-one, naphtholbenzein, 1 4h-naphthalenone, 4-4-hydroxy-1-naphthalenyl phenylmethylene, 1 4h-naphthalenone,4-4-hydroxy-1-naphthalenyl phenylmethylene, 1 4h-naphthalenone, 4-.alpha.-4-hydroxy-1-naphthyl benzylidene
PubChem CID 5941340
IUPAC Name (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one
SMILES C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Quantity 10 g
Formula Weight 374.44
Chemical Name or Material α-Naphtholbenzein, 98%
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.