accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNHN

CAS RN 1450-85-7

IUPAC Name: 1,2-dihydropyrimidine-2-thione
Synonyms: 2-(1H)-pyrimidinthione
Molecular Weight (g/mol): 112.15
Molecular Formula: C4H4N2S
InChi Key: HBCQSNAFLVXVAY-UHFFFAOYSA-N
SMILES: S=C1NC=CC=N1
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (5)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)