Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptopyrimidine 98.0+%, TCI America™
SDP

Catalog No. M093725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M093725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M093725G Supplier TCI America Supplier No. M093725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1450-85-7
Molecular Formula C4H4N2S
Molecular Weight (g/mol) 112.15
MDL Number MFCD00006073
InChI Key HBCQSNAFLVXVAY-UHFFFAOYSA-N
Synonym 2-mercaptopyrimidine, pyrimidine-2-thiol, 2-pyrimidinethiol, 2-thiopyrimidine, pyrimidine-2 1h-thione, 2 1h-pyrimidinethione, pyrimidinethione, 2-pyrimidinethione, 2-pyrimidinethiol 9ci, 2-1h-pyrimidinthione
PubChem CID 1550489
ChEBI CHEBI:53576
IUPAC Name 1H-pyrimidine-2-thione
SMILES C1=CNC(=S)N=C1

Specifications

Quantity 25 g
Formula Weight 112.15
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercaptopyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.