accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CCH3SOOO

CAS RN 145022-45-3

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium methanesulfonate
Synonyms: 1-ethyl-3-methylimidazolium mesylate
Molecular Weight (g/mol): 206.26
Molecular Formula: C7H14N2O3S
InChi Key: IXLWEDFOKSJYBD-UHFFFAOYSA-M
SMILES: CS([O-])(=O)=O.CCN1C=C[N+](C)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)