Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-methylimidazolium Methanesulfonate 98.0+%, TCI America™
SDP

Catalog No. E07555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07555G 5 g
E075525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07555G Supplier TCI America Supplier No. E07555G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 145022-45-3
Molecular Formula C7H14N2O3S
Molecular Weight (g/mol) 206.26
MDL Number MFCD06798171
InChI Key IXLWEDFOKSJYBD-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium methanesulfonate, 1-ethyl-3-methylimidazolium mesylate, basionics'st 35, basionics™ st 35, acmc-1c0dz, dsstox_cid_29075, dsstox_rid_83294, dsstox_gsid_49219, 1-ethyl-3-methyl imidazolium methyl sulfonate, 1-ethyl-3-methylimidazolium me-thanesulfonate
PubChem CID 16211849
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;methanesulfonate
SMILES CCN1C=C[N+](=C1)C.CS(=O)(=O)[O-]

Specifications

Quantity 5 g
UN Number 3265
Formula Weight 206.26
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-methylimidazolium Methanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.