CAS RN 14528-64-4
IUPAC Name:
dipotassium 5-nitro-2-(sulfonatooxy)benzen-1-olate dihydrate
Synonyms:
dipotassium 5-nitro-2-(sulfonatooxy)benzenolate dihydrate
Molecular Weight (g/mol):
347.38
Molecular Formula:
C6H7K2NO9S
InChi Key:
ZIAQNWGTQNXFHV-UHFFFAOYSA-L
SMILES:
O.O.[K+].[K+].[O-]C1=CC(=CC=C1OS([O-])(=O)=O)[N+]([O-])=O