accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14529-53-4

IUPAC Name: 2-ethoxypyridine
Synonyms: 2-ethoxypyridine
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: LISKAOIANGDBTB-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results