Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxypyridine 98.0+%, TCI America™
SDP

Catalog No. E08705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08705G 5 g
E087025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08705G Supplier TCI America Supplier No. E08705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14529-53-4
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00023464
InChI Key LISKAOIANGDBTB-UHFFFAOYSA-N
Synonym pyridine, 2-ethoxy, 2-ethoxy pyridine, 2-ethoxy-pyridine, zlchem 304, pubchem1126, acmc-209cun, ethyl 2-pyridinyl ether, ksc497c7d
PubChem CID 84499
IUPAC Name 2-ethoxypyridine
SMILES CCOC1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 157°C
Quantity 5 g
UN Number 1993
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.