CAS RN 14538-51-3
IUPAC Name:
N-[3-(hydroxyamino)-2,3-dimethylbutan-2-yl]hydroxylamine; sulfuric acid
Synonyms:
N-[3-(hydroxyamino)-2,3-dimethylbutan-2-yl]hydroxylamine; sulfuric acid
Molecular Weight (g/mol):
246.28
Molecular Formula:
C6H18N2O6S
InChi Key:
IUVKJBVHRBBGOL-UHFFFAOYSA-N
SMILES:
OS(O)(=O)=O.CC(C)(NO)C(C)(C)NO
1
–
1
of
1
results
1
2,3-Bis(hydroxyamino)-2,3-dimethylbutane Sulfate (Pract.), 85%, Thermo Scientific™
CAS: 14538-51-3 Molecular Formula: C6H16N2O2.H2SO4 Synonym: N,N'-Dihydroxy-2,3-dimethyl-2,3-butanediamine sulfate