accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNH2OONHSOOCH2SOOO

CAS RN 14541-90-3

IUPAC Name: sodium 1-amino-4-{[2-(ethenesulfonyl)phenyl]amino}-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
Synonyms: sodium 1-amino-4-{[2-(ethenesulfonyl)phenyl]amino}-9,10-dioxoanthracene-2-sulfonate
Molecular Weight (g/mol): 506.48
Molecular Formula: C22H15N2NaO7S2
InChi Key: ODADSOUDUYEAHK-UHFFFAOYSA-M
SMILES: [Na+].NC1=C2C(=O)C3=CC=CC=C3C(=O)C2=C(NC2=CC=CC=C2S(=O)(=O)C=C)C=C1S([O-])(=O)=O
Color 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)