accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOSOOFFF

CAS RN 145412-54-0

IUPAC Name: dicyclohexylboranyl trifluoromethanesulfonate
Synonyms: dicyclohexylboranyl trifluoromethanesulfonate
Molecular Weight (g/mol): 326.18
Molecular Formula: C13H22BF3O3S
InChi Key: FBVDLYBQGQLANQ-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OB(C1CCCCC1)C1CCCCC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)