Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicyclohexyl(trifluoromethanesulfonyloxy)borane 98.0+%, TCI America™
SDP

Catalog No. D29435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29435G 5 g
D29431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29435G Supplier TCI America Supplier No. D29435G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 145412-54-0
Molecular Formula C13H22BF3O3S
Molecular Weight (g/mol) 326.181
MDL Number MFCD06797094
InChI Key FBVDLYBQGQLANQ-UHFFFAOYSA-N
Synonym Dicyclohexylboron Triflate, Trifluoromethanesulfonic Acid Dicyclohexylboryl Ester
PubChem CID 10958346
IUPAC Name dicyclohexylboranyl trifluoromethanesulfonate
SMILES B(C1CCCCC1)(C2CCCCC2)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
UN Number 1759
Formula Weight 326.18
Percent Purity ≥97.0% (T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Dicyclohexyl(trifluoromethanesulfonyloxy)borane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.