accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1455-05-6

IUPAC Name: 2-chloroethyl chlorophosphonochloridate
Synonyms: 2-chloroethyl chlorophosphonochloridate
Molecular Weight (g/mol): 197.38
Molecular Formula: C2H4Cl3O2P
InChi Key: YRTAUZXAHQPFJV-UHFFFAOYSA-N
SMILES: ClCCOP(Cl)(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results