CAS RN 145543-82-4
IUPAC Name:
2-bromo-3-butylthiophene
Synonyms:
2-bromo-3-butylthiophene
Molecular Weight (g/mol):
219.14
Molecular Formula:
C8H11BrS
InChi Key:
KNBLITSBUDZSJW-UHFFFAOYSA-N
SMILES:
CCCCC1=C(Br)SC=C1
1
–
1
of
1
results
1
2-Bromo-3-butylthiophene 98.0+%, TCI America™
CAS: 145543-82-4 Molecular Formula: C8H11BrS Molecular Weight (g/mol): 219.14 MDL Number: MFCD16619253 InChI Key: KNBLITSBUDZSJW-UHFFFAOYSA-N PubChem CID: 15354891 IUPAC Name: 2-bromo-3-butylthiophene SMILES: CCCCC1=C(SC=C1)Br